Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stl
Entrez Gene ID
37619
Gene Name
stall
Gene Aliases
BcDNA:GM08694, CG3622, Dmel\CG3622, Dmel_CG3622, fs(2)A16
Chromosome Location
Chr.2R: 22807860 - 22815993 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086408
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103953.3 NP_001097423.1 6 - 7 2015 68
NM_001259540.1 NP_001246469.1 6 - 7 2523 68

View Full Product Details