Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4ad1
Entrez Gene ID
35821
Gene Name
CG2110-PA
Gene Aliases
4ad1, BEST:LD02646, CG2110, Cyp4AD1, Dmel\CG2110, Dmel_CG2110
Chromosome Location
Chr.2R: 8439395 - 8442806 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033292
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273876.1 NP_001260805.1 4 - 5 563 60
NM_136536.3 NP_610380.1 6 - 7 968 60
GenBank mRNA AY061058.1 - 6 - 7 941 60
BT133029.1 - 4 - 5 563 60

View Full Product Details