The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169803.2 | NP_001163274.1 | 5 - 6 | 1130 | 67 |
NM_001169805.2 | NP_001163276.1 | 4 - 5 | 957 | 67 | |
NM_001169807.2 | NP_001163278.2 | 4 - 5 | 1243 | 67 | |
NM_137970.5 | NP_611814.3 | 5 - 6 | 988 | 67 | |
NM_166616.3 | NP_726350.1 | 4 - 5 | 1243 | 67 | |
NM_166617.2 | NP_726351.1 | 2 - 3 | 377 | 67 | |
NM_166618.3 | NP_726352.1 | 3 - 4 | 726 | 67 | |
GenBank mRNA | BT011336.1 | - | 4 - 5 | 1207 | 67 |
BT021326.1 | - | 5 - 6 | 984 | 67 | |
BT133111.1 | - | 3 - 4 | 726 | 67 |