The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103889.2 | NP_001097359.1 | 3 - 4 | 3164 | 80 |
NM_001299609.1 | NP_001286538.1 | 4 - 5 | 2581 | 80 | |
NM_079051.4 | NP_523775.2 | 3 - 4 | 2641 | 80 |