Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snap29
- Entrez Gene ID
-
37774
- Gene Name
- Synaptosomal-associated protein 29kDa
- Gene Aliases
-
CG11173, Dmel\CG11173, Dmel_CG11173, MENE (2R)-E, MENE(2R)-E, SNAP-29, SNAP29, Ubisnap, dSNAP-29, ubisnap, usnp
- Chromosome Location
-
Chr.2R: 23843483 - 23844763 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034913
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079107.4 |
NP_523831.1 |
1 - 2 |
380 |
59 |
GenBank mRNA |
AY061069.1 |
- |
1 - 2 |
338 |
59 |
View Full Product Details