Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Awh
Entrez Gene ID
38451
Gene Name
Arrowhead
Gene Aliases
CG1072, Dmel\CG1072, Dmel_CG1072, awh, l(3)63Ea, l(3)63Ea/Awh, l(3)64Ea, l(3)SH9, l63Ea
Chromosome Location
Chr.3L: 3846206 - 3861692 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013751
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274450.1 NP_001261379.1 5 - 6 1301 65
NM_001274451.1 NP_001261380.1 5 - 6 1289 65
NM_079183.3 NP_523907.2 5 - 6 1236 65
NM_168042.2 NP_728906.1 5 - 6 1301 65
GenBank mRNA BT025230.1 - 5 - 6 1296 65

View Full Product Details