Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab19
Entrez Gene ID
38930
Gene Name
CG7062-PA
Gene Aliases
AAF50452, CG7062, DRABR3, DRabRP3, DmRab19, Dmel\CG7062, Dmel_CG7062, O18339, Rab-RP3, Rab-r3, Rab19/RP3, Rab19s, rab19
Chromosome Location
Chr.3L: 8297018 - 8298506 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015793
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274646.1 NP_001261575.1 3 - 4 422 117
NM_079246.4 NP_523970.1 2 - 3 393 117
GenBank mRNA AY071400.1 - 2 - 3 386 117
D84348.1 - 2 - 3 374 117

View Full Product Details