Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
row
Entrez Gene ID
36685
Gene Name
relative of woc
Gene Aliases
CG8092, Dmel\CG8092, Dmel_CG8092, Row, anon-WO0118547.194
Chromosome Location
Chr.2R: 15206527 - 15213634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033998
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274063.2 NP_001260992.1 5 - 6 3138 90
NM_137175.4 NP_611019.2 5 - 6 3138 90
GenBank mRNA BT016036.1 - 5 - 6 3117 90

View Full Product Details