Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mamo
Entrez Gene ID
32353
Gene Name
maternal gene required for meiosis
Gene Aliases
CG11071, CG11072, CG11075, CG11082, CG32606, CG32611, CG34346, CG44132, CG45477, CR44656, Dmel\CG45477, Dmel_CG45477, lincRNA.990
Chromosome Location
Chr.X: 13846116 - 13990527 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267033
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103508.4 NP_001096978.1 6 - 7 4072 58
NM_001201660.3 NP_001188589.1 4 - 5 2982 58
NM_001258732.2 NP_001245661.1 6 - 7 3843 58
NM_001258733.1 NP_001245662.1 5 - 6 2967 58
NM_001272610.2 NP_001259539.1 6 - 7 3857 58
NM_132704.4 NP_572932.2 6 - 7 3857 58
NM_167387.5 NP_727743.3 6 - 7 4072 58
GenBank mRNA BT133085.1 - 4 - 5 2462 58

View Full Product Details