Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Prosbeta5R1
- Entrez Gene ID
-
37690
- Gene Name
- Proteasome beta5 subunit-related 1
- Gene Aliases
-
CG9868, Dmel\CG9868, Dmel_CG9868, Prosbeta5R, beta5R1
- Chromosome Location
-
Chr.2R: 23308654 - 23309816 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034842
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137932.2 |
NP_611776.1 |
1 - 2 |
238 |
96 |
View Full Product Details