Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ItgaPS5
- Entrez Gene ID
-
37732
- Gene Name
- Integrin alphaPS5 subunit
- Gene Aliases
-
CG5372, CT17068, Dmel\CG5372, Dmel_CG5372, ItgalphaPS5, alpha-PS5, alphaPS5
- Chromosome Location
-
Chr.2R: 23556672 - 23560263 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034880
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137964.2 |
NP_611808.2 |
2 - 3 |
1092 |
73 |
View Full Product Details