Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Iml1
- Entrez Gene ID
-
38176
- Gene Name
- Increased minichromosome loss 1
- Gene Aliases
-
CG12090, Dmel\CG12090, Dmel_CG12090, SEA1, iml1
- Chromosome Location
-
Chr.3L: 1523414 - 1534034 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035227
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139361.2 |
NP_647618.1 |
10 - 11 |
3065 |
91 |
View Full Product Details