Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xmas
Entrez Gene ID
44271
Gene Name
CG46386-PA
Gene Aliases
CG32561, CG32562, CG46386, CG8919, Dmel\CG46386, Dmel_CG46386, Xmas-2, l(1)15Ea, m(1)3, ms(1)3, ms3, xmas-1, xmas-2
Chromosome Location
Chr.X: 17151231 - 17159306 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0286809
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370033.1 NP_001356948.1 4 - 5 5162 74
NM_080034.3 NP_524773.2 2 - 3 1153 74

View Full Product Details