Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- plu
- Entrez Gene ID
-
44848
- Gene Name
- plutonium
- Gene Aliases
-
CG9183, Dmel\CG9183, Dmel_CG9183, PLU, Plu
- Chromosome Location
-
Chr.2R: 20262958 - 20263684 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003114
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057335.3 |
NP_476683.1 |
2 - 3 |
445 |
57 |
View Full Product Details