Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elk
Entrez Gene ID
37047
Gene Name
Eag-like K[+] channel
Gene Aliases
CG5076, Dmel\CG5076, Dmel_CG5076, dmELK, elk
Chromosome Location
Chr.2R: 17873939 - 17926695 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011589
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299624.1 NP_001286553.1 13 - 14 1893 64
NM_057661.3 NP_477009.1 13 - 14 1896 64

View Full Product Details