Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sb
Entrez Gene ID
41958
Gene Name
Stubble
Gene Aliases
CG4316, Dmel\CG4316, Dmel_CG4316, SP56, Sb-sbd, c-SP56, cSP56, sbd, st-sb
Chromosome Location
Chr.3R: 16116161 - 16145284 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003319
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300426.1 NP_001287355.1 5 - 6 1971 113
NM_057361.4 NP_476709.1 5 - 6 1727 113

View Full Product Details