Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir85a
- Entrez Gene ID
-
41052
- Gene Name
- Ionotropic receptor 85a
- Gene Aliases
-
CG11775, CT32618, DmelIR85a, Dmel\CG11775, Dmel_CG11775, IR85a
- Chromosome Location
-
Chr.3R: 8794684 - 8796966 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037630
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141576.2 |
NP_649833.1 |
2 - 3 |
1387 |
75 |
View Full Product Details