Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ScsbetaA
Entrez Gene ID
41067
Gene Name
Succinyl-coenzyme A synthetase beta subunit, ADP-forming
Gene Aliases
A-Sbeta, CG11963, CGI 1963, Dmel\CG11963, Dmel_CG11963, FBgn0037643, SkAP, Skap, skap
Chromosome Location
Chr.3R: 8938724 - 8943027 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037643
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104243.3 NP_001097713.1 5 - 6 818 62
NM_001202268.3 NP_001189197.2 6 - 7 790 62
NM_001202269.2 NP_001189198.2 5 - 6 818 62
NM_141589.4 NP_649846.2 6 - 7 790 62

View Full Product Details