Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl5
Entrez Gene ID
41438
Gene Name
methuselah-like 5
Gene Aliases
CG6965, Dmel\CG6965, Dmel_CG6965, Gia, Mth-5, Mthl5, anon-WO0170980.70, anon-WO0170980.71
Chromosome Location
Chr.3R: 11883938 - 11887168 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037960
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300362.1 NP_001287291.1 3 - 4 883 87
NM_141869.3 NP_650126.2 3 - 4 883 87
GenBank mRNA BT100276.1 - 3 - 4 880 87

View Full Product Details