Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wge
Entrez Gene ID
42687
Gene Name
winged eye
Gene Aliases
CG17621, CG31151, CG4902, Dmel\CG31151, Dmel_CG31151
Chromosome Location
Chr.3R: 22710883 - 22721150 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051151
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043281.2 NP_001036746.1 5 - 6 3567 84
NM_001275918.1 NP_001262847.1 5 - 6 3603 84
NM_170037.2 NP_732790.3 5 - 6 3600 84
NM_170038.2 NP_732791.2 4 - 5 3450 84
GenBank mRNA AB190214.1 - 5 - 6 2973 84
BT099586.1 - 5 - 6 3554 84

View Full Product Details