Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dmrt99B
Entrez Gene ID
43495
Gene Name
doublesex-Mab related 99B
Gene Aliases
CG15504, Dmel\CG15504, Dmel_CG15504
Chromosome Location
Chr.3R: 29692591 - 29697025 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039683
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079825.2 NP_524549.1 1 - 2 368 59
GenBank mRNA BT022192.1 - 1 - 2 344 59
BT023707.1 - 1 - 2 344 59

View Full Product Details