Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppi1
Entrez Gene ID
43581
Gene Name
Protein phosphatase 1c interacting protein 1
Gene Aliases
CG2164, CG31025, Dmel\CG31025, Dmel_CG31025, anon-WO0140519.94, ppi1
Chromosome Location
Chr.3R: 30125633 - 30129119 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051025
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170466.2 NP_733345.2 1 - 2 731 75
NM_170467.2 NP_733346.1 1 - 2 731 75
GenBank mRNA AY119510.1 - 1 - 2 714 75

View Full Product Details