Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MtnA
Entrez Gene ID
41202
Gene Name
Metallothionein A
Gene Aliases
BcDNA:GH18460, CG9470, Dmel\CG9470, Dmel_CG9470, DrosMtn, MTA, MTN, Mtn, anon-WO0153538.42, anon-WO0153538.44, anon-WO0153538.49, mtnA
Chromosome Location
Chr.3R: 9783407 - 9784370 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002868
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275509.1 NP_001262438.1 2 - 2 276 77
NM_079575.2 NP_524299.1 2 - 2 276 77
GenBank mRNA BT001431.1 - 2 - 2 332 77
K02314.1 - 2 - 2 274 77

View Full Product Details