Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cep43
Entrez Gene ID
415215
Gene Name
centrosomal protein 43
Gene Aliases
ccr6a, fgfr1op, zgc:86782
Chromosome Location
Chr.20: 30578966 - 30610172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002125.1 NP_001002125.1 16 - 17 1357 106
XM_021468391.1 XP_021324066.1 18 - 19 1529 106
GenBank mRNA BC071450.1 - 16 - 17 1357 106

View Full Product Details