Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
id2b
Entrez Gene ID
321364
Gene Name
inhibitor of DNA binding 2b
Gene Aliases
hm:zeh0152, id2l, zgc:73382
Chromosome Location
Chr.20: 29784920 - 29787273 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199541.1 NP_955835.1 1 - 2 386 112
XM_005160630.2 XP_005160687.1 1 - 2 449 112
GenBank mRNA BC059690.1 - 1 - 2 386 112
BC097036.1 - 1 - 2 398 112
BC165495.1 - 1 - 2 373 112

View Full Product Details