Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gata1a
Entrez Gene ID
30481
Gene Name
GATA binding protein 1a
Gene Aliases
gata1, gta1, sb:eu551, vlt, zg1
Chromosome Location
Chr.11: 37854 - 44803 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131234.2 NP_571309.2 5 - 6 1076 118
GenBank mRNA BC124483.1 - 5 - 6 1004 118
BC164788.1 - 4 - 5 1012 118
U18311.1 - 5 - 6 118

View Full Product Details