Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crispld1b
Entrez Gene ID
393442
Gene Name
cysteine-rich secretory protein LCCL domain containing 1b
Gene Aliases
crispld1, zgc:63636
Chromosome Location
Chr.2: 30379627 - 30396476 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200470.1 NP_956764.1 5 - 6 866 89
XM_009298567.3 XP_009296842.1 5 - 6 889 89
GenBank mRNA BC055176.1 - 5 - 6 866 89

View Full Product Details