Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
samsn1b
Entrez Gene ID
450018
Gene Name
SAM domain, SH3 domain and nuclear localisation signals 1b
Gene Aliases
zgc:103427
Chromosome Location
Chr.10: 38396151 - 38428095 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006039.2 NP_001006039.2 6 - 7 830 64
XM_005169164.4 XP_005169221.1 14 - 15 1780 64
XM_009305561.3 XP_009303836.1 14 - 15 1777 64
GenBank mRNA BC083374.1 - 6 - 7 803 64
EH436758.1 - 6 - 7 840 64

View Full Product Details