Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mxe
Entrez Gene ID
337663
Gene Name
myxovirus (influenza virus) resistance E
Gene Aliases
wu:fj93f01, zgc:111895
Chromosome Location
Chr.9: 32824572 - 32839888 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182867.1 NP_878287.1 2 - 3 360 126
XM_005167664.4 XP_005167721.2 3 - 4 492 126
GenBank mRNA AJ544827.1 - 2 - 3 360 126
BC095587.1 - 3 - 4 402 126

View Full Product Details