Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhrs3b
Entrez Gene ID
450050
Gene Name
dehydrogenase/reductase (SDR family) member 3b
Gene Aliases
ik:tdsubc_1f3, tdsubc_1f3, xx:tdsubc_1f3, zgc:101705
Chromosome Location
Chr.11: 28590733 - 28614608 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006070.1 NP_001006070.1 1 - 2 613 74
GenBank mRNA BC083252.1 - 1 - 2 613 74
BC164070.1 - 1 - 2 231 74

View Full Product Details