Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
drd2a
Entrez Gene ID
282557
Gene Name
dopamine receptor D2a
Gene Aliases
-
Chromosome Location
Chr.15: 22045778 - 22074372 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183068.1 NP_898891.1 6 - 7 1155 62
XM_005157501.4 XP_005157558.1 7 - 8 1603 62
XM_009291617.3 XP_009289892.1 6 - 7 1519 62
GenBank mRNA AY183456.1 - 6 - 7 1155 62

View Full Product Details