The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007438.1 | NP_001007439.1 | 6 - 7 | 531 | 59 |
GenBank mRNA | BC085405.1 | - | 6 - 7 | 531 | 59 |