Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cep44
Entrez Gene ID
767769
Gene Name
centrosomal protein 44
Gene Aliases
zgc:153894
Chromosome Location
Chr.1: 38362309 - 38374799 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076736.2 NP_001070204.2 4 - 5 486 78
XM_021475843.1 XP_021331518.1 4 - 5 525 78
GenBank mRNA BC124486.1 - 4 - 5 468 78

View Full Product Details