Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tagln3b
Entrez Gene ID
797861
Gene Name
transgelin 3b
Gene Aliases
fc80d07, tagln3, wu:fc80d07, zgc:66052
Chromosome Location
Chr.24: 21917211 - 21923873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199720.1 NP_956014.1 2 - 3 387 84
GenBank mRNA BC062382.1 - 2 - 3 387 84

View Full Product Details