Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan15
Entrez Gene ID
100000710
Gene Name
tetraspanin 15
Gene Aliases
im:7155083, zgc:110375
Chromosome Location
Chr.13: 22926117 - 22961591 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017773.1 NP_001017773.1 1 - 2 197 126
GenBank mRNA BC092907.1 - 1 - 2 197 126
BC152126.1 - 1 - 2 106 126

View Full Product Details