Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rmnd1
Entrez Gene ID
553810
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
zgc:113022
Chromosome Location
Chr.17: 7795561 - 7819002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020772.1 NP_001018608.1 5 - 6 713 68
XM_009292818.3 XP_009291093.1 5 - 6 953 68
XM_017351638.2 XP_017207127.1 5 - 6 774 68
XM_021467167.1 XP_021322842.1 5 - 6 642 68
GenBank mRNA BC095858.1 - 5 - 6 713 68

View Full Product Details