Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bmpr2b
Entrez Gene ID
555655
Gene Name
bone morphogenetic protein receptor, type II b (serine/threonine kinase)
Gene Aliases
-
Chromosome Location
Chr.9: 12959760 - 13111163 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039807.1 NP_001034896.1 6 - 7 1667 63
GenBank mRNA BC163200.1 - 6 - 7 1525 63
DQ002899.1 - 6 - 7 1667 63

View Full Product Details