Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
impdh2
Entrez Gene ID
317745
Gene Name
IMP (inosine 5'-monophosphate) dehydrogenase 2
Gene Aliases
IMPD 2, IMPDH 2, cb635, wu:fb64g02, wu:fc43f09
Chromosome Location
Chr.8: 26059563 - 26079140 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201464.1 NP_958872.1 6 - 7 786 76
XM_021478184.1 XP_021333859.1 6 - 7 897 76
GenBank mRNA BC046905.1 - 6 - 7 786 76

View Full Product Details