Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pkp3a
Entrez Gene ID
368397
Gene Name
plakophilin 3a
Gene Aliases
pkp3, wu:fa97f09, wu:fk69a12, zgc:136656
Chromosome Location
Chr.25: 22151271 - 22191728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045280.1 NP_001038745.1 11 - 12 2596 88
GenBank mRNA BC116608.1 - 11 - 12 2596 88
BC155136.1 - 11 - 12 2615 88

View Full Product Details