Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mttp
Entrez Gene ID
406207
Gene Name
microsomal triglyceride transfer protein
Gene Aliases
mtp, stl, wu:fd36b01, zgc:111876, zgc:136353
Chromosome Location
Chr.1: 12107680 - 12126541 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212970.2 NP_998135.2 7 - 8 1086 87
GenBank mRNA AJ428850.1 - 7 - 8 979 87
BC135020.1 - 7 - 8 1063 87

View Full Product Details