Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tlcd2
Entrez Gene ID
559420
Gene Name
TLC domain containing 2
Gene Aliases
zgc:175098
Chromosome Location
Chr.15: 25399507 - 25435021 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110382.1 NP_001103852.1 2 - 3 621 141
GenBank mRNA BC154828.1 - 2 - 3 621 141

View Full Product Details