Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kctd16a
Entrez Gene ID
567207
Gene Name
potassium channel tetramerization domain containing 16a
Gene Aliases
lrl1
Chromosome Location
Chr.14: 115021 - 115938 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037238.1 NP_001032315.1 1 - 1 554 79
GenBank mRNA AY763407.1 - 1 - 1 554 79
BC163117.1 - 1 - 1 550 79
BC163138.1 - 1 - 1 550 79

View Full Product Details