Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pus3
Entrez Gene ID
337487
Gene Name
pseudouridylate synthase 3
Gene Aliases
fj78b12, wu:fj78b12, zgc:56653
Chromosome Location
Chr.24: 36317639 - 36329660 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200067.1 NP_956361.1 4 - 5 922 90
GenBank mRNA BC049516.1 - 4 - 5 922 90

View Full Product Details