Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tlr2
Entrez Gene ID
403125
Gene Name
toll-like receptor 2
Gene Aliases
-
Chromosome Location
Chr.1: 8983491 - 8989759 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212812.1 NP_997977.1 1 - 1 2254 103
XM_005170853.4 XP_005170910.1 2 - 2 2921 103
GenBank mRNA AY388399.1 - 1 - 1 2254 103

View Full Product Details