Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tpk1
Entrez Gene ID
450054
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
zgc:101685
Chromosome Location
Chr.24: 18286427 - 18444729 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006074.1 NP_001006074.1 9 - 9 1051 86
XM_005162615.4 XP_005162672.1 9 - 9 86
GenBank mRNA BC083247.1 - 9 - 9 1051 86

View Full Product Details