Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mul1b
Entrez Gene ID
100004181
Gene Name
mitochondrial E3 ubiquitin protein ligase 1b
Gene Aliases
im:7146383, zgc:165594
Chromosome Location
Chr.23: 37286509 - 37293429 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098760.1 NP_001092230.1 3 - 4 253 78
XM_005166206.4 XP_005166263.1 3 - 4 246 78
GenBank mRNA BC142887.1 - 3 - 4 253 78

View Full Product Details