Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fstl1b
Entrez Gene ID
324309
Gene Name
follistatin-like 1b
Gene Aliases
fa95c03, fc25c04, wu:fa95c03, wu:fc25c04
Chromosome Location
Chr.9: 9736397 - 9842197 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039621.1 NP_001034710.1 8 - 9 803 83
XM_009304495.3 XP_009302770.1 8 - 9 848 83
GenBank mRNA DQ317971.1 - 8 - 9 803 83

View Full Product Details