Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tkfc
Entrez Gene ID
793926
Gene Name
triokinase/FMN cyclase
Gene Aliases
dak, zgc:153296
Chromosome Location
Chr.7: 19738679 - 19760223 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076630.1 NP_001070098.1 5 - 6 722 92
GenBank mRNA BC124306.1 - 5 - 6 722 92

View Full Product Details