Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifnphi2
Entrez Gene ID
100126802
Gene Name
interferon phi 2
Gene Aliases
ifn2
Chromosome Location
Chr.3: 22352552 - 22356568 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111082.1 NP_001104552.1 1 - 2 130 63

View Full Product Details