Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lhx4
Entrez Gene ID
571943
Gene Name
LIM homeobox 4
Gene Aliases
si:dkeyp-35f11.3
Chromosome Location
Chr.8: 14453928 - 14488693 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122973.1 NP_001116445.1 2 - 3 252 63
XM_021478301.1 XP_021333976.1 2 - 3 286 63
XM_021478302.1 XP_021333977.1 2 - 3 278 63
GenBank mRNA BC129374.1 - 2 - 3 421 63

View Full Product Details